Assay Details
Target Gene Details
Entrez Gene ID: | 11686 |
Gene Name: | arachidonate 12-lipoxygenase, 12R type |
Gene Aliases: |
Aloxe2, e-LOX2 |
Location: |
Chr.11:69157072-69169791 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alox12b | NM_009659.2 | NP_033789.1 | ||
| AF059251.1 | AAC79681.1 | |||
| BC113149.1 | AAI13150.1 | |||
| Y14334.1 | CAA74714.1 |
More Information
Set Membership: |
|

Genomic Map