Assay Details
Target Gene Details
Entrez Gene ID: | 216991 |
Gene Name: | ArfGAP with dual PH domains 2 |
Gene Aliases: |
Centa2 |
Location: |
Chr.11:80153896-80178827 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adap2 | NM_172133.1 | NP_742145.1 | ||
| AK050079.1 | BAE20669.1 | |||
| AK050398.1 | BAE20671.1 | |||
| AK053152.1 | ||||
| AK139005.1 | BAE23859.1 | |||
| AK140864.1 | BAE24501.1 | |||
| BC027165.1 | AAH27165.1 |

Genomic Map