Assay Details
Target Gene Details
Entrez Gene ID: | 66179 |
Gene Name: | 2-oxoglutarate and iron-dependent oxygenase domain containing 3 |
Gene Aliases: |
1110031I02Rik |
Location: |
Chr.11:121177593-121204648 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ogfod3 | NM_025402.2 | NP_079678.1 | ||
| AK004005.1 | BAB23119.1 | |||
| AK007883.1 | ||||
| AK145938.1 | BAE26768.1 | |||
| AK168300.1 | BAE40241.1 | |||
| BC024349.1 | AAH24349.1 |
More Information
Set Membership: |
|

Genomic Map