Assay Details
Target Gene Details
Entrez Gene ID: | 217265 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 5 |
Gene Aliases: |
ABC13, B930033A02Rik, mKIAA1888 |
Location: |
Chr.11:110269369-110337724 on Build GRCm38 |
Assay Gene Location: | Within Exon 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca5 | NM_147219.2 | 36 | 4900 | NP_671752.2 |
| AB097675.1 | 35 | 4650 | BAC66658.1 | |
| AF491842.1 | 36 | 4859 | AAM90895.1 | |
| AK129463.1 | 14 | 1553 | BAC98273.1 | |
| AK138604.1 | 36 | 4717 | BAE23713.1 |
More Information
Set Membership: |
|

Genomic Map