Assay Details
Target Gene Details
Entrez Gene ID: | 109934 |
Gene Name: | active BCR-related gene |
Gene Aliases: |
- |
Location: |
Chr.11:76416732-76577739 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abr | NM_001291186.1 | NP_001278115.1 | ||
| NM_198018.2 | NP_932135.1 | |||
| NM_198894.2 | NP_942597.1 | |||
| NM_198895.2 | NP_942598.1 | |||
| AK134993.1 | BAE22375.1 | |||
| AK219799.1 | ||||
| BC056385.1 | AAH56385.1 | |||
| BC058708.1 | AAH58708.1 | |||
| BC059064.1 | AAH59064.1 |
More Information
Set Membership: |
|

Genomic Map