Assay Details
Target Gene Details
Entrez Gene ID: | 76025 |
Gene Name: | calcium activated nucleotidase 1 |
Gene Aliases: |
5830420C20Rik, Apy1h, D11Bwg0554e, Entpd8, SCAN-1, Shapy |
Location: |
Chr.11:118406289-118419134 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cant1 | NM_001025617.2 | 4 | 983 | NP_001020788.1 |
| NM_001025618.2 | 4 | 941 | NP_001020789.1 | |
| NM_001267591.1 | 3 | 784 | NP_001254520.1 | |
| NM_001267592.1 | 5 | 1094 | NP_001254521.1 | |
| NM_029502.3 | 4 | 1135 | NP_083778.2 | |
| AK006565.1 | 5 | 945 | BAB24655.1 | |
| AK049879.1 | 4 | 877 | BAC33968.1 | |
| AK081118.1 | 4 | 1040 | BAC38139.1 | |
| AK085059.1 | 3 | 2231 | BAC39351.1 | |
| AK166384.1 | 3 | 2145 | ||
| BC020003.1 | 3 | 783 | AAH20003.1 | |
| CA320376.1 |
More Information
Set Membership: |
|

Genomic Map