Assay Details
Target Gene Details
Entrez Gene ID: | 23801 |
Gene Name: | arachidonate lipoxygenase 3 |
Gene Aliases: |
e-LOX-3, eLOX-3 |
Location: |
Chr.11:69125674-69149115 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aloxe3 | NM_011786.2 | NP_035916.2 | ||
| BC116258.1 | ||||
| BC116259.1 | ||||
| Y14695.1 | CAB46101.1 |
More Information
Set Membership: |
|

Genomic Map