Assay Details
Target Gene Details
Entrez Gene ID: | 76184 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 6 |
Gene Aliases: |
6330565N06Rik |
Location: |
Chr.11:110174638-110251776 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 31 - Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca6 | NM_147218.2 | NP_671751.2 | ||
| AF498361.1 | AAM90907.1 | |||
| BC132417.1 | ||||
| BC132419.1 |
More Information
Set Membership: |
|

Genomic Map