Assay Details
Target Gene Details
Entrez Gene ID: | 11927 |
Gene Name: | ATX1 (antioxidant protein 1) homolog 1 (yeast) |
Gene Aliases: |
AI256639, Atx1 |
Location: |
Chr.11:55446643-55461138 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atox1 | NM_009720.2 | NP_033850.1 | ||
| AF004591.1 | AAB61465.1 | |||
| AK002432.1 | BAB22098.1 | |||
| BC027632.1 | AAH27632.1 |

Genomic Map