Assay Details
Target Gene Details
Entrez Gene ID: | 268294 |
Gene Name: | zinc finger and BTB domain containing 24 |
Gene Aliases: |
BC055367, Bif1, Bsg1, ZNF450 |
Location: |
Chr.10:41450356-41465582 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zbtb24 | NM_001277229.1 | NP_001264158.1 | ||
| NM_001277230.1 | NP_001264159.1 | |||
| NM_153398.3 | NP_700447.2 | |||
| NR_102360.1 | ||||
| AF263010.2 | AAK91811.2 | |||
| AK054097.1 | BAE20695.1 | |||
| AK146986.1 | BAE27589.1 | |||
| AK154467.1 | ||||
| AK169260.1 | ||||
| AY210402.1 | AAO42998.1 | |||
| BC050933.1 | AAH50933.1 | |||
| BC055367.1 | AAH55367.1 |
More Information
Set Membership: |
|

Genomic Map