Assay Details
Target Gene Details
Entrez Gene ID: | 216739 |
Gene Name: | acyl-CoA synthetase long-chain family member 6 |
Gene Aliases: |
A330035H04Rik, AW050338, Facl6, LACS, Lacsl, mKIAA0837 |
Location: |
Chr.11:54303824-54364756 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsl6 | NM_001033597.1 | NP_001028769.1 | ||
| NM_001033598.1 | NP_001028770.1 | |||
| NM_001033599.1 | NP_001028771.1 | |||
| NM_144823.4 | NP_659072.3 | |||
| AB073974.1 | ||||
| AK032479.1 | BAC27892.1 | |||
| AK039380.1 | BAC30332.1 | |||
| AK147283.1 | BAE27819.1 | |||
| AK147421.1 | BAE27902.1 | |||
| AY167035.1 | AAO38689.1 | |||
| AY786360.1 | AAW33883.1 | |||
| AY786361.1 | AAW33884.1 | |||
| AY786362.1 | AAW33885.1 | |||
| AY786363.1 | AAW33886.1 | |||
| BC016114.1 | AAH16114.1 | |||
| BC022959.1 |
More Information
Set Membership: |
|

Genomic Map