Assay Details
Target Gene Details
Entrez Gene ID: | 104184 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
AI035728, Bh, Bmh |
Location: |
Chr.11:76945656-76987389 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blmh | NM_178645.4 | 1 | 162 | NP_848760.1 |
| AK029274.1 | 1 | 81 | ||
| AK035228.1 | BAC28989.1 | |||
| AK040799.1 | BAC30706.1 | |||
| AK049461.1 | 1 | 74 | ||
| AK051441.1 | 1 | 61 | BAC34639.1 | |
| AK053872.1 | BAC35568.1 | |||
| AK150438.1 | 1 | 61 | BAE29560.1 | |
| AK150896.1 | ||||
| AK167325.1 | BAE39427.1 | |||
| AK197894.1 | ||||
| AK199579.1 | ||||
| BC024090.1 | AAH24090.1 | |||
| BC027037.1 | AAH27037.1 | |||
| BC027362.1 | AAH27362.1 | |||
| BC027403.1 | 1 | 72 | AAH27403.1 | |
| BY087076.1 | 1 | 162 |
More Information
Set Membership: |
|

Genomic Map