Assay Details
Target Gene Details
Entrez Gene ID: | 104184 |
Gene Name: | bleomycin hydrolase |
Gene Aliases: |
AI035728, Bh, Bmh |
Location: |
Chr.11:76945656-76987389 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blmh | NM_178645.4 | 2 | 297 | NP_848760.1 |
| AK029274.1 | 2 | 216 | ||
| AK035228.1 | 2 | 171 | BAC28989.1 | |
| AK040799.1 | 2 | 187 | BAC30706.1 | |
| AK049461.1 | 2 | 209 | ||
| AK051441.1 | 2 | 196 | BAC34639.1 | |
| AK053872.1 | 2 | 177 | BAC35568.1 | |
| AK150438.1 | 2 | 196 | BAE29560.1 | |
| AK150896.1 | 2 | 177 | ||
| AK167325.1 | 2 | 177 | BAE39427.1 | |
| AK197894.1 | 2 | 101 | ||
| AK199579.1 | 2 | 101 | ||
| BC024090.1 | 2 | 182 | AAH24090.1 | |
| BC027037.1 | 2 | 153 | AAH27037.1 | |
| BC027362.1 | 2 | 186 | AAH27362.1 | |
| BC027403.1 | 2 | 207 | AAH27403.1 | |
| BY087076.1 | 2 | 297 |
More Information
Set Membership: |
|

Genomic Map