Assay Details
Target Gene Details
Entrez Gene ID: | 12351 |
Gene Name: | carbonic anhydrase 4 |
Gene Aliases: |
AW456718, Ca4 |
Location: |
Chr.11:84957754-84966054 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Car4 | NM_007607.2 | 1 | 137 | NP_031633.1 |
AK154754.1 | 1 | 103 | BAE32805.1 | |
AK165399.1 | 1 | 65 | ||
BC012704.1 | AAH12704.1 |
