Assay Details
Target Gene Details
Entrez Gene ID: | 11370 |
Gene Name: | acyl-Coenzyme A dehydrogenase, very long chain |
Gene Aliases: |
vlcad |
Location: |
Chr.11:70010183-70015428 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acadvl | NM_017366.3 | 10 | 1087 | NP_059062.1 |
| AF017176.1 | 7 | 735 | AAC31642.1 | |
| AK076037.1 | 10 | 1048 | ||
| AK146334.1 | 5 | 560 | BAE27089.1 | |
| AK189988.1 | 1 | 61 | ||
| AK208302.1 | 1 | 61 | ||
| BC026559.1 | 10 | 1066 | AAH26559.1 | |
| Z71189.1 | 10 | 1008 | CAA94919.2 |
More Information
Set Membership: |
|

Genomic Map