Assay Details
Target Gene Details
Entrez Gene ID: | 11736 |
Gene Name: | ankyrin repeat and FYVE domain containing 1 |
Gene Aliases: |
Ankhzn, ZFYVE14, mKIAA1255 |
Location: |
Chr.11:72690002-72772146 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankfy1 | NM_009671.5 | 3 | 280 | NP_033801.4 |
| AB011370.2 | 3 | 278 | BAA24980.2 | |
| AB098329.1 | 3 | 278 | BAC67211.1 | |
| AK134008.1 | 3 | 277 | ||
| BC139231.1 | 3 | 291 | ||
| BC145583.1 | 3 | 291 | ||
| BQ550282.1 | 3 | 285 |
More Information
Set Membership: |
|

Genomic Map