Assay Details
Target Gene Details
Entrez Gene ID: | 66884 |
Gene Name: | amyloid beta precursor protein (cytoplasmic tail) binding protein 2 |
Gene Aliases: |
1300003O07Rik, AI465480, PAT1 |
Location: |
Chr.11:85191308-85235120 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Appbp2 | NM_025825.3 | NP_080101.1 | ||
| AK004888.1 | ||||
| AK005440.1 | BAB24034.1 | |||
| AK133853.1 | ||||
| AK177656.1 | ||||
| AK182121.1 | ||||
| AK207679.1 | ||||
| AK207768.1 | ||||
| AK217461.1 | ||||
| BC018442.1 | AAH18442.1 | |||
| BC019152.1 | AAH19152.1 |

Genomic Map