Assay Details
Target Gene Details
Entrez Gene ID: | 327957 |
Gene Name: | SLP adaptor and CSK interacting membrane protein |
Gene Aliases: |
A430084P05Rik |
Location: |
Chr.11:70790932-70812561 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Scimp | NM_001045526.2 | NP_001038991.1 | ||
AK138818.1 | BAE23788.1 | |||
BC147177.1 | ||||
BC147178.1 |
