Assay Details
Target Gene Details
Entrez Gene ID: | 217258 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 8a |
Gene Aliases: |
- |
Location: |
Chr.11:110025634-110097583 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 32 - Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca8a | NM_153145.4 | NP_694785.3 | ||
| AB255881.1 | ||||
| AB255882.1 | ||||
| AF498360.1 | AAM90906.1 | |||
| AK031843.1 | BAC27576.1 | |||
| AY732492.1 | AAU81985.1 | |||
| BC026496.1 | AAH26496.1 | |||
| BC060032.1 | AAH60032.1 |
More Information
Set Membership: |
|

Genomic Map