Assay Details
Target Gene Details
Entrez Gene ID: | 11370 |
Gene Name: | acyl-Coenzyme A dehydrogenase, very long chain |
Gene Aliases: |
vlcad |
Location: |
Chr.11:70010183-70015428 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acadvl | NM_017366.3 | NP_059062.1 | ||
| AF017176.1 | AAC31642.1 | |||
| AK076037.1 | ||||
| AU067294.1 | ||||
| BC026559.1 | AAH26559.1 | |||
| Z71189.1 | CAA94919.2 |
More Information
Set Membership: |
|

Genomic Map