Assay Details
Target Gene Details
Entrez Gene ID: | 544817 |
Gene Name: | Rho GTPase activating protein 27 |
Gene Aliases: |
2310069I04Rik, 5730442P18Rik, Gm21552, Sh3d20 |
Location: |
Chr.11:103331484-103363692 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Arhgap27 | NM_001205236.1 | NP_001192165.1 | ||
| NM_133715.5 | NP_598476.2 | |||
| AK046763.1 | BAE20660.1 | |||
| AK143916.1 | BAE25601.1 | |||
| AK154926.1 | ||||
| BC096671.1 | AAH96671.1 |
More Information
Set Membership: |
|

Genomic Map