Assay Details
Target Gene Details
Entrez Gene ID: | 53334 |
Gene Name: | golgi SNAP receptor complex member 1 |
Gene Aliases: |
AI414660, AI426320, BB145494, GOS-28, GOSRI, GS28 |
Location: |
Chr.11:76726602-76763622 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gosr1 | NM_016810.3 | 3 | 195 | NP_058090.2 |
| AF079901.1 | 3 | 218 | AAC32189.1 | |
| AK133836.1 | 3 | 205 | BAE21873.1 | |
| AK134515.1 | 3 | 198 | BAE22167.1 | |
| AK147799.1 | 3 | 219 | BAE28147.1 | |
| AK154972.1 | 3 | 221 | BAE32964.1 | |
| AK161386.1 | 3 | 209 | BAE36365.1 | |
| BC008542.1 | 3 | 195 | AAH08542.1 | |
| BC051661.1 | 3 | 214 | AAH51661.1 |
More Information
Set Membership: |
|

Genomic Map