Assay Details
Target Gene Details
Entrez Gene ID: | 12409 |
Gene Name: | carbonyl reductase 2 |
Gene Aliases: |
MLCR |
Location: |
Chr.11:120729485-120732025 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cbr2 | NM_007621.2 | NP_031647.1 | ||
| AK081816.1 | BAC38340.1 | |||
| BC010758.1 | AAH10758.1 | |||
| D26123.1 | BAA05120.1 | |||
| X07411.1 | CAA30309.1 |
More Information
Set Membership: |
|

Genomic Map