Assay Details
Target Gene Details
Entrez Gene ID: | 216859 |
Gene Name: | ArfGAP with coiled-coil, ankyrin repeat and PH domains 1 |
Gene Aliases: |
Centb1 |
Location: |
Chr.11:69881567-69895539 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acap1 | NM_153788.3 | NP_722483.2 | ||
| AK133473.1 | 6 | 1452 | ||
| AK138736.1 | ||||
| AK154210.1 | ||||
| AK154449.1 | BAE32594.1 | |||
| AK155001.1 | ||||
| BC031462.1 | AAH31462.1 | |||
| BC037481.1 | AAH37481.1 |

Genomic Map