Assay Details
Target Gene Details
Entrez Gene ID: | 14104 |
Gene Name: | fatty acid synthase |
Gene Aliases: |
A630082H08Rik, FAS |
Location: |
Chr.11:120805958-120824547 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 40 - Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fasn | NM_007988.3 | NP_032014.3 | ||
| AF127033.1 | AAG02285.1 | |||
| AK080374.1 | BAC37895.1 | |||
| AK147214.1 | BAE27771.1 | |||
| AK147694.1 | ||||
| BC046513.1 | AAH46513.1 | |||
| BC059850.1 | AAH59850.1 | |||
| X13135.1 | CAA31525.1 |

Genomic Map