Assay Details
Target Gene Details
Entrez Gene ID: | 16834 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
6030445I15, Ldlb, mKIAA1381 |
Location: |
Chr.11:113649178-113669331 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cog1 | NM_013581.3 | NP_038609.3 | ||
| AF109377.2 | AAD13780.2 | |||
| AK031510.1 | ||||
| AK034636.2 | BAE43288.1 | |||
| AK152464.1 | BAE31241.1 | |||
| AK158507.1 | BAE34539.1 | |||
| AK159497.1 | BAE35130.1 | |||
| AK162971.1 | BAE37141.1 | |||
| AK219755.1 | ||||
| AK220387.1 | BAD90252.1 | |||
| BC049541.1 | AAH49541.1 | |||
| BC063056.1 | AAH63056.1 |

Genomic Map