Assay Details
Target Gene Details
Entrez Gene ID: | 16834 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
6030445I15, Ldlb, mKIAA1381 |
Location: |
Chr.11:113649178-113669331 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cog1 | NM_013581.3 | 8 | 2207 | NP_038609.3 |
AF109377.2 | 8 | 2190 | AAD13780.2 | |
AK031510.1 | ||||
AK034636.2 | 8 | 2212 | BAE43288.1 | |
AK152464.1 | 8 | 2198 | BAE31241.1 | |
AK158507.1 | 8 | 2569 | BAE34539.1 | |
AK159497.1 | 8 | 2209 | BAE35130.1 | |
AK162971.1 | 8 | 2208 | BAE37141.1 | |
AK171574.1 | 8 | 2201 | BAE42534.1 | |
AK177359.1 | ||||
AK185855.1 | ||||
AK220387.1 | 7 | 2651 | BAD90252.1 | |
BC049541.1 | 2 | 657 | AAH49541.1 | |
BC063056.1 | 8 | 2189 | AAH63056.1 |
