Assay Details
Target Gene Details
Entrez Gene ID: | 16834 |
Gene Name: | component of oligomeric golgi complex 1 |
Gene Aliases: |
6030445I15, Ldlb, mKIAA1381 |
Location: |
Chr.11:113649178-113669331 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cog1 | NM_013581.3 | 9 | 2364 | NP_038609.3 |
| AF109377.2 | 9 | 2347 | AAD13780.2 | |
| AK031510.1 | 8 | 1782 | ||
| AK034636.2 | 9 | 2369 | BAE43288.1 | |
| AK152464.1 | 9 | 2355 | BAE31241.1 | |
| AK158507.1 | 9 | 2726 | BAE34539.1 | |
| AK159497.1 | 9 | 2366 | BAE35130.1 | |
| AK162971.1 | 9 | 2365 | BAE37141.1 | |
| AK171574.1 | 9 | 2358 | BAE42534.1 | |
| AK177622.1 | 1 | 133 | ||
| AK187486.1 | 1 | 133 | ||
| AK189116.1 | 1 | 133 | ||
| AK217864.1 | 1 | 133 | ||
| AK220387.1 | 8 | 2808 | BAD90252.1 | |
| BC049541.1 | 3 | 814 | AAH49541.1 | |
| BC063056.1 | 9 | 2346 | AAH63056.1 |

Genomic Map