Assay Details
Target Gene Details
Entrez Gene ID: | 260302 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 3 |
Gene Aliases: |
C230037M19Rik, mKIAA0154 |
Location: |
Chr.11:115584255-115603917 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gga3 | NM_001252067.1 | 14 | 1623 | NP_001238996.1 |
NM_173048.3 | 15 | 1857 | NP_766636.2 | |
AK031086.1 | 15 | 1829 | BAC27246.1 | |
AK082327.1 | 14 | 1587 | ||
AK122212.2 | 15 | 1826 | BAC65494.2 | |
AK132183.1 | 15 | 1825 | BAE21019.1 | |
AK147882.1 | 15 | 1825 |
