Assay Details
Target Gene Details
Entrez Gene ID: | 216527 |
Gene Name: | cerebral cavernous malformation 2 |
Gene Aliases: |
BC029157 |
Location: |
Chr.11:6546887-6596761 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccm2 | NM_001190343.1 | 3 | 316 | NP_001177272.1 |
| NM_001190344.1 | 4 | 402 | NP_001177273.1 | |
| NM_146014.3 | 4 | 490 | NP_666126.1 | |
| AK155145.1 | 4 | 464 | BAE33075.1 | |
| AK166131.1 | BAE38589.1 | |||
| AK169727.1 | 4 | 404 | ||
| AK189178.1 | ||||
| AK197140.1 | 1 | 89 | ||
| AK207564.1 | 1 | 89 | ||
| AY442689.1 | 4 | 377 | AAR29082.1 | |
| BC029157.1 | 4 | 421 | AAH29157.1 |

Genomic Map