Assay Details
Target Gene Details
Entrez Gene ID: | 216527 |
Gene Name: | cerebral cavernous malformation 2 |
Gene Aliases: |
BC029157 |
Location: |
Chr.11:6546887-6596761 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccm2 | NM_001190343.1 | NP_001177272.1 | ||
| NM_001190344.1 | NP_001177273.1 | |||
| NM_146014.3 | NP_666126.1 | |||
| AK155145.1 | BAE33075.1 | |||
| AK166131.1 | BAE38589.1 | |||
| AK169727.1 | ||||
| AK189544.1 | ||||
| AY442689.1 | AAR29082.1 | |||
| BC029157.1 | AAH29157.1 |

Genomic Map