Assay Details
Target Gene Details
Entrez Gene ID: | 13385 |
Gene Name: | discs, large homolog 4 (Drosophila) |
Gene Aliases: |
Dlgh4, PSD-95, PSD95, SAP90, SAP90A |
Location: |
Chr.11:70018455-70045532 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlg4 | NM_001109752.1 | 12 | 1730 | NP_001103222.1 |
| NM_007864.3 | 12 | 1739 | NP_031890.1 | |
| BC014807.1 | 12 | 1475 | AAH14807.1 | |
| BQ959940.1 | ||||
| D50621.1 | 12 | 1481 | BAA09297.1 |
More Information
Set Membership: |
|

Genomic Map