Assay Details
Target Gene Details
Entrez Gene ID: | 68626 |
Gene Name: | elaC homolog 2 (E. coli) |
Gene Aliases: |
1110017O07Rik, D11Wsu80e, Hpc2 |
Location: |
Chr.11:64979023-65002076 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Elac2 | NM_023479.2 | NP_075968.2 | ||
| AF308696.2 | AAG24918.2 | |||
| AK003759.1 | BAB22981.2 | |||
| AK004136.1 | BAB23185.1 | |||
| AK156730.1 | BAE33824.1 | |||
| AK178054.1 | ||||
| AK185294.1 | ||||
| AK186537.1 | ||||
| AK194083.1 | ||||
| AK194247.1 | ||||
| AK194420.1 | ||||
| BC048235.1 | AAH48235.1 |
More Information
Set Membership: |
|

Genomic Map