Assay Details
Target Gene Details
Entrez Gene ID: | 260302 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 3 |
Gene Aliases: |
C230037M19Rik, mKIAA0154 |
Location: |
Chr.11:115584255-115603917 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Gga3 | NM_001252067.1 | 5 | 417 | NP_001238996.1 |
NM_173048.3 | 5 | 417 | NP_766636.2 | |
AK031086.1 | 5 | 389 | BAC27246.1 | |
AK082327.1 | 5 | 381 | ||
AK122212.2 | 5 | 386 | BAC65494.2 | |
AK132183.1 | 5 | 385 | BAE21019.1 | |
AK147882.1 | 5 | 386 | ||
AK213811.1 | ||||
AK216206.1 | ||||
BY268727.1 | ||||
BY296723.1 |
