Assay Details
Target Gene Details
Entrez Gene ID: | 14025 |
Gene Name: | B cell CLL/lymphoma 11A (zinc finger protein) |
Gene Aliases: |
2810047E18Rik, BCL-11A, Ctip1, D930021L15Rik, Evi9, Evi9a, Evi9b, Evi9c, mKIAA1809 |
Location: |
Chr.11:24076564-24173558 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Bcl11a |
