Assay Details
Target Gene Details
Entrez Gene ID: | 12777 |
Gene Name: | chemokine (C-C motif) receptor 10 |
Gene Aliases: |
Cmkbr9, Gpr2 |
Location: |
Chr.11:101172922-101175695 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ccr10 | NM_007721.4 | NP_031747.2 | ||
AF215982.1 | AAF63710.1 | |||
AK090044.1 | BAC41062.1 | |||
BC117043.1 | ||||
BC120542.1 |
Target Gene Details
Entrez Gene ID: | 53321 |
Gene Name: | contactin associated protein-like 1 |
Gene Aliases: |
AI841080, Caspr, NCP1, Nrxn4, p190, shm |
Location: |
Chr.11:101174629-101190727 on Build GRCm38 |
Assay Gene Location: |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cntnap1 |
