Assay Details
Target Gene Details
Entrez Gene ID: | 216848 |
Gene Name: | chromodomain helicase DNA binding protein 3 |
Gene Aliases: |
2600010P09Rik, AF020312, Chd7, Prp7, Prp9-1 |
Location: |
Chr.11:69343273-69369426 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 39 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Chd3 | NM_146019.4 | NP_666131.3 | ||
AK011183.1 | ||||
BC030435.1 | AAH30435.1 | |||
BC055484.1 | AAH55484.1 |
