Assay Details
Target Gene Details
Entrez Gene ID: | 70207 |
Gene Name: | translational activator of mitochondrially encoded cytochrome c oxidase I |
Gene Aliases: |
2310066I18Rik, Ccdc44 |
Location: |
Chr.11:106066039-106073612 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Taco1 | NM_027346.1 | NP_081622.1 | ||
AK010072.1 | BAB26682.1 | |||
AK047078.1 | BAC32953.1 | |||
AK140143.1 | BAE24253.1 | |||
AK147942.1 | ||||
AK167963.1 | BAE39960.1 | |||
BC029182.1 | AAH29182.1 |
