Assay Details
Target Gene Details
Entrez Gene ID: | 12467 |
Gene Name: | chaperonin containing Tcp1, subunit 6b (zeta) |
Gene Aliases: |
CCT-zeta-2, CCTzeta-2, Cctz-2, TCP-1-zeta-2 |
Location: |
Chr.11:82719248-82764321 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cct6b | NM_001291242.1 | NP_001278171.1 | ||
| NM_009839.3 | NP_033969.2 | |||
| AK076639.1 | BAC36429.1 | |||
| BC100457.1 | AAI00458.1 | |||
| Z50192.1 | CAA90574.1 |
More Information
Set Membership: |
|

Genomic Map