Assay Details
Target Gene Details
Entrez Gene ID: | 73321 |
Gene Name: | RIKEN cDNA 1700042O10 gene |
Gene Aliases: |
- |
Location: |
Chr.11:11868123-11885321 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 1700042O10Rik | NR_045178.1 | |||
| AK006690.1 |
Target Gene Details
Entrez Gene ID: | 13195 |
Gene Name: | dopa decarboxylase |
Gene Aliases: |
Aadc |
Location: |
Chr.11:11814101-11898144 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddc | NM_001190448.1 | 2 | 241 | NP_001177377.1 |
| NM_016672.4 | 2 | 179 | NP_057881.1 | |
| AF012881.1 | 2 | 130 | AAB84011.1 | |
| AF012882.1 | 1 | 109 | AAB84012.1 | |
| AF071068.1 | 1 | 112 | AAC25566.1 | |
| AK011834.1 | 2 | 173 | ||
| AK149141.1 | 2 | 173 | BAE28750.1 | |
| AK160769.1 | 2 | 173 | BAE35999.1 | |
| BY129740.1 | 2 | 180 |
More Information
Set Membership: |
|

Genomic Map