Assay Details
Target Gene Details
Entrez Gene ID: | 380711 |
Gene Name: | RAP1 GTPase activating protein 2 |
Gene Aliases: |
AU067654, Garnl4, Gm1561, mKIAA1039 |
Location: |
Chr.11:74383483-74610910 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap1gap2 | NM_001015046.2 | 14 | 1276 | NP_001015046.1 |
| AK038834.1 | 4 | 339 | BAE20542.1 | |
| AK147290.1 | 15 | 1258 | ||
| BC107388.2 | 5 | 482 | AAI07389.1 |
More Information
Set Membership: |
|

Genomic Map