Assay Details
Target Gene Details
Entrez Gene ID: | 380711 |
Gene Name: | RAP1 GTPase activating protein 2 |
Gene Aliases: |
AU067654, Garnl4, Gm1561, mKIAA1039 |
Location: |
Chr.11:74383483-74610910 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rap1gap2 | NM_001015046.2 | NP_001015046.1 | ||
| AK147290.1 | ||||
| AW457490.1 | ||||
| BY239088.1 | ||||
| CF743736.1 | ||||
| CJ106176.1 | ||||
| CJ111553.1 |
More Information
Set Membership: |
|

Genomic Map