Assay Details
Target Gene Details
Entrez Gene ID: | 216963 |
Gene Name: | G protein-coupled receptor kinase-interactor 1 |
Gene Aliases: |
Cat-1, p95Cat |
Location: |
Chr.11:77493412-77507775 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Git1 | NM_001004144.1 | 18 | 2120 | NP_001004144.1 |
AK050701.1 | 5 | 438 | BAE20687.1 | |
BC079870.1 | 18 | 2120 | AAH79870.1 |
