Assay Details
Target Gene Details
Entrez Gene ID: | 13056 |
Gene Name: | cytochrome b-561 |
Gene Aliases: |
- |
Location: |
Chr.11:105933702-105945114 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyb561 | NM_007805.4 | NP_031831.2 | ||
| AK014395.2 | BAB29321.1 | |||
| AK028780.1 | BAC26117.1 | |||
| AK158978.1 | BAE34753.1 | |||
| AK169711.1 | BAE41322.1 | |||
| BC006732.1 | AAH06732.1 | |||
| U16297.1 | AAA65643.1 |
More Information
Set Membership: |
|

Genomic Map