Assay Details
Target Gene Details
Entrez Gene ID: | 15218 |
Gene Name: | forkhead box N1 |
Gene Aliases: |
D11Bhm185e, Fkh19, HFH-11, Hfh11, Whn, nu, nude |
Location: |
Chr.11:78357577-78386922 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Foxn1 | NM_001277290.1 | 6 | 1024 | NP_001264219.1 |
NM_008238.2 | 6 | 1027 | NP_032264.1 | |
BC108980.1 | 5 | 912 | AAI08981.1 | |
BC108981.1 | 5 | 915 | AAI08982.1 | |
X81593.1 | 6 | 977 | CAA57279.1 |
