Assay Details
Target Gene Details
Entrez Gene ID: | 14615 |
Gene Name: | gap junction protein, gamma 1 |
Gene Aliases: |
C130009G16Rik, Cnx45, Cx45, Gja-7, Gja7 |
Location: |
Chr.11:102799472-102819686 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gjc1 | NM_001159383.1 | NP_001152855.1 | ||
| AY390396.1 | ||||
| BC071230.1 | AAH71230.1 |
More Information
Set Membership: |
|

Genomic Map