Assay Details
Target Gene Details
Entrez Gene ID: | 78394 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 52 |
Gene Aliases: |
2700029C06Rik, ROK1 |
Location: |
Chr.11:83942090-83963088 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx52 | NM_030096.2 | NP_084372.2 | ||
| AK080767.1 | BAC38014.1 | |||
| AK133545.1 | BAE21716.1 | |||
| AK141200.1 | ||||
| AK166789.1 | BAE39019.1 | |||
| AK195121.1 | ||||
| BC029094.1 | AAH29094.1 |

Genomic Map