Assay Details
Target Gene Details
Entrez Gene ID: | 13808 |
Gene Name: | enolase 3, beta muscle |
Gene Aliases: |
Eno-3, MSE |
Location: |
Chr.11:70654763-70662513 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eno3 | NM_001136062.2 | 7 | 682 | NP_001129534.1 |
| NM_001276285.1 | 7 | 694 | NP_001263214.1 | |
| NM_007933.3 | 7 | 794 | NP_031959.1 | |
| AK002485.1 | 7 | 654 | BAB22137.1 | |
| AK193383.1 | ||||
| BC013460.1 | 6 | 666 | AAH13460.1 | |
| CT010193.1 | 6 | 588 | CAJ18401.1 | |
| M20745.1 | 4 | 414 | AAA37554.1 | |
| X57747.1 | 7 | 645 | CAA40913.1 | |
| X62667.1 | 6 | 638 | CAA44540.1 |
More Information
Set Membership: |
|

Genomic Map