Assay Details
Target Gene Details
Entrez Gene ID: | 17846 |
Gene Name: | COMM domain containing 1 |
Gene Aliases: |
AI256843, Murr1, U2/Mu |
Location: |
Chr.11:22899728-22982843 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Commd1 | NM_144514.2 | 1 | 106 | NP_653097.2 |
| AB104816.1 | BAC57942.1 | |||
| AK082909.1 | 1 | 82 | ||
| AK133507.1 | 1 | 82 | BAE21693.1 | |
| BC030052.1 | AAH30052.1 | |||
| BC051210.1 | 1 | 67 | AAH51210.1 | |
| BC132083.1 | 1 | 95 | ||
| BC132085.1 | 1 | 96 | ||
| BY337798.1 | 1 | 107 | ||
| D85430.1 | 1 | 76 |

Genomic Map