Assay Details
Target Gene Details
Entrez Gene ID: | 52513 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 56 |
Gene Aliases: |
2600001H07Rik, D11Ertd619e, Noh61 |
Location: |
Chr.11:6257545-6267729 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ddx56 | NM_026538.3 | NP_080814.1 | ||
AK011136.1 | BAB27426.1 | |||
AK076047.1 | BAC36145.1 | |||
AK076975.1 | BAC36543.1 | |||
AK077514.1 | BAC36839.1 | |||
AK077953.1 | ||||
AK145628.1 | BAE26549.1 | |||
AK159793.1 | BAE35375.1 | |||
AK169654.1 | BAE41278.1 | |||
AK185833.1 | ||||
BC018291.1 | AAH18291.1 |
